missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, 99.999%, (trace metal basis), for analysis, anhydrous, powder

Product Code. 10364112 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
20 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
20g
This item is not returnable. View return policy

Product Code. 10364112

Brand: Thermo Scientific undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis, Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 194.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula AlCl3
Fieser 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15
Merck Index 15,333
Solubility Information Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform
Formula Weight 133.34
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass Bottle
Specific Gravity 2.44
Density 2.4400g/mL
Chemical Name or Material Aluminum chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.