missing translation for 'onlineSavingsMsg'
Learn More
Aluminum chloride, 99%, extra pure, anhydrous, granules, Thermo Scientific Chemicals Product Code.: 10636782

Aluminum chloride, 99%, extra pure, anhydrous, granules, Thermo Scientific Chemicals

Product Code. 10636782
1 kg, Glass bottle
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10636782

Brand: Thermo Scientific Acros 195780010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminumShow MoreShow Less
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 194.0°C
Color Yellow
Physical Form Granules
Quantity 1 kg
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula AlCl3
Fieser 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15
Merck Index 15, 333
Solubility Information Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform
Formula Weight 133.34
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 2.44
Density 2.4400g/mL
Chemical Name or Material Aluminum chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466127 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number195780010,195785000
Lot NumberA0464601 Certificate TypeCertificate of Analysis Date17/07/2024 Catalog Number195780010,195785000
Lot NumberA0458670 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number195780010,195785000
Lot NumberA0443733 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number195780010,195785000
Lot NumberA0401262 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number195780010,195785000
compliance-icons
Product Identifier
  • Aluminium chloride
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum chloride, 99%, extra pure, anhydrous, granules, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.