missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid

Specifications
Specifications
Quantity | 2.5 kg |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | AlCl3·6H2O |
Fieser | 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15 |
Merck Index | 15,333 |
Solubility Information | Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol,ether,glycerol and propylene |
IUPAC Name | aluminum(3+) hexahydrate trichloride |
Formula Weight | 241.43 |
Percent Purity | 99% |
Grade | Pure |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468313 | Certificate of Analysis | 31/01/2025 | 217470025,217470100,217470010,217475000 |
A0467853 | Certificate of Analysis | 30/01/2025 | 217470025,217470100,217470010,217475000 |
A0467097 | Certificate of Analysis | 31/10/2024 | 217470025,217470100,217470010,217475000 |
A0467098 | Certificate of Analysis | 29/10/2024 | 217470025,217470100,217470010,217475000 |
A0466859 | Certificate of Analysis | 15/10/2024 | 217470025,217470100,217470010,217475000 |
Safety and Handling

Product Identifier
- Aluminium chloride hexahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction