missing translation for 'onlineSavingsMsg'
Learn More
Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals Product Code.: 11914181

Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals

Product Code. 11914181
10 kg, Plastic drum
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 11914181

Brand: Thermo Scientific Acros 217470100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Quantity 10 kg
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula AlCl3·6H2O
Fieser 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15
Merck Index 15,333
Solubility Information Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol, ether, glycerol and propylene
IUPAC Name aluminum(3+) hexahydrate trichloride
Formula Weight 241.43
Percent Purity 99%
Grade Pure
Packaging Plastic drum
Specific Gravity 2.39
Density 2.3900g/mL
Chemical Name or Material Aluminum chloride hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468313 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number217470025,217470100,217470010,217475000
Lot NumberA0467853 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number217470025,217470100,217470010,217475000
Lot NumberA0467097 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number217470025,217470100,217470010,217475000
Lot NumberA0467098 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number217470025,217470100,217470010,217475000
Lot NumberA0466859 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number217470025,217470100,217470010,217475000
compliance-icons
Product Identifier
  • Aluminium chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.