Learn More
Aluminum isopropoxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
£47.30 - £635.00
Chemical Identifiers
CAS | 555-31-7 |
---|---|
Molecular Formula | C9H21AlO3 |
Molecular Weight (g/mol) | 204.25 |
MDL Number | MFCD00008870 |
InChI Key | SMZOGRDCAXLAAR-UHFFFAOYSA-N |
Synonym | aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate |
PubChem CID | 11143 |
IUPAC Name | aluminum;propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10195422
|
Thermo Scientific Acros
212230100 |
10 g | Glass bottle |
£47.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10546413
|
Thermo Scientific Acros
212230500 |
50 g | Glass bottle |
£153.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10716433
|
Thermo Scientific Acros
212232500 |
250 g | Glass Bottle |
£635.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Lewis acidChemical Identifiers
555-31-7 | |
204.25 | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
11143 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
C9H21AlO3 | |
MFCD00008870 | |
aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
aluminum;propan-2-olate |
Specifications
555-31-7 | |
White | |
Authentic | |
C9H21AlO3 | |
MFCD00008870 | |
15,342 | |
Solubility in water: reacts. Other solubilities: freely soluble in ethanol | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] | |
204.25 | |
204.25 | |
Trace Metal Basis | |
Aluminum isopropoxide |
129.0°C to 136.0°C | |
125.0°C to 130.0°C (38.0 mmHg) | |
99.99% | |
[(CH3)2CHO]3Al | |
01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16 | |
aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
aluminum;propan-2-olate | |
11143 | |
99.99% | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Flammable solid.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing d
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-090-8
RUO – Research Use Only