missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Nitrate Nonahydrate, 98+%, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
Specifications
Specifications
| Melting Point | 73°C |
| pH | 2.5 to 3.5 |
| Physical Form | Solid |
| Quantity | 1 kg |
| Formula Weight | 375.13g/mol |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Aluminium nitrate nonahydrate
- Warning
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction