Learn More
Aluminum nitrate nonahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
£44.92 - £219.09
Chemical Identifiers
CAS | 7784-27-2 |
---|---|
Molecular Formula | AlN3O9·9H2O |
Molecular Weight (g/mol) | 375.13 |
InChI Key | SWCIQHXIXUMHKA-UHFFFAOYSA-N |
Synonym | aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronicShow More |
PubChem CID | 24567 |
IUPAC Name | aluminum;trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11954271
|
Thermo Scientific Acros
212240100 |
10 g | Glass bottle |
£44.92
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964271
|
Thermo Scientific Acros
212241000 |
100 g | Glass bottle |
£219.09
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7784-27-2 | |
375.13 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronicShow More | |
aluminum;trinitrate;nonahydrate |
Specifications
73.0°C | |
White | |
6.5 to 7.9% (Al) (Complexometry) | |
Al(NO3)3·9H2O | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 | |
99.999% | |
Aluminum nitrate nonahydrate |
7784-27-2 | |
Crystals | |
AlN3O9·9H2O | |
15,347 | |
Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.13 | |
375.13 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Immediately call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
- Aluminium nitrate nonahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Aluminum nitrate nonahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.