missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum nitrate nonahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 73.0°C |
pH | 2.5 to 3.5 (5% soln. at 20°C) |
Physical Form | Crystals |
Quantity | 10 kg |
Assay Percent Range | 99+% |
Molecular Formula | AlH18N3O18 |
Linear Formula | Al(NO3)3·9H2O |
Solubility Information | Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine |
Molecular Weight (g/mol) | 375.129 |
Formula Weight | 375.13 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'