Learn More
Aluminum oxide, 99.7+%, extra pure, Thermo Scientific Chemicals
£21.85 - £126.47
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10441641
|
Thermo Scientific Acros
215702500 |
250 g | Plastic bottle |
£21.85
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042820
|
Thermo Scientific Acros
215700010 |
1 kg | Plastic bottle |
£38.99
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137713
|
Thermo Scientific Acros
215700050 |
5 kg | Plastic bottle |
£126.47
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundumShow More | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
2977.0°C | |
1344-28-1 | |
Powder | |
Al2O3 | |
MFCD00003424 | |
15, 350 | |
Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic, solvents, difficultly soluble in mineral acids | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Extra Pure | |
3.9700g/mL |
2030.0°C | |
White | |
99.7+% | |
AI2O3 | |
01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
≥99.7% | |
3.97 | |
Aluminum oxide, Extra Pure |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.