missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, for chromatography, basic, Brockmann I, 50-200 μm, 90A, Thermo Scientific Chemicals

Product Code. 16534234
Click to view available options
Quantity:
1 kg

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2977.0°C
Melting Point 2030.0°C
CAS 1344-28-1
Quantity 1 kg
Molecular Formula Al2O3
Linear Formula Al2O3
MDL Number MFCD00003424
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
Formula Weight 101.96
Grade Chromatography
Specific Gravity 3.97
Density 3.9700 g/mL
Chemical Name or Material Aluminum oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.