missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals
89.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 2980.0°C |
Melting Point | 2050.0°C |
Color | White |
pH | 6.5 to 7.5 (5% aq. suspension, stirred) |
Physical Form | Fine Powder |
Quantity | 2.5 kg |
Linear Formula | Al2O3 |
Fieser | 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9 |
Merck Index | 15, 350 |
Solubility Information | Solubility in water: insoluble |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'