missing translation for 'onlineSavingsMsg'
Learn More
Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 10120362

Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 10120362
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10120362

Brand: Thermo Scientific Acros 217485000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlKO8S2·12H2O
Molecular Weight (g/mol) 474.39
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrateShow MoreShow Less
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Melting Point 92.0°C
Color White
pH 3 to 3.5 (10% soln. at 20°C)
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99.5% min. (Complexometry)
Linear Formula AlK(SO4)2·12H2O
Merck Index 15, 354
Solubility Information Solubility in water: 140g/L (20°C). Other solubilities: freely soluble in glycerol, insoluble in alcohol
Formula Weight 474.39
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Aluminum potassium sulfate dodecahydrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464452 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number217485000,217480025,217480010
Lot NumberA0455274 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number217485000,217480025,217480010
Lot NumberA0450100 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog Number217485000,217480025,217480010
Lot NumberA0403308 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number217485000,217480025,217480010
Lot NumberA0394995 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number217485000,217480025,217480010
Product Identifier
  • Aluminium potassium sulfate dodecahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.