missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10765321
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum Potassium Sulfate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Weight (g/mol) 474.37
MDL Number 149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
IUPAC Name aluminium(3+) potassium dodecahydrate disulfate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 92°C
Color White
pH 3.0 to 3.5
Physical Form Solid
Quantity 1 kg
Molecular Formula AlH24KO20S2
Formula Weight 474.39g/mol
Packaging Plastic powder jar
Product Identifier
  • Aluminium potassium sulfate dodecahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'