missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 195805000
27.65 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Aluminum standard solution | |
1 mg/ml Al in 0.5N HNO3 | |
Plastic bottle | |
Yellow | |
MFCD00134029 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 | |
AAS | |
-3.0°C | |
1.02 | |
Powder |
990 to 1010μg/mL of Al(3+) in 2 to 5% HNO3 | |
7784-27-2, 7697-37-2, 7732-18-5 | |
500 mL | |
AlH18N3O18 | |
15, 347 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.129 | |
375.13 | |
1.0200g/mL | |
101.0°C | |
Soluble in water |
RUO – Research Use Only