missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

£36.88 - £337.68

Chemical Identifiers

CAS 10043-01-3
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.13
MDL Number MFCD00003423
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
PubChem CID 24850
ChEBI CHEBI:74768
IUPAC Name dialuminum;trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10542601
View Documents
Thermo Scientific Acros
192430050
5 g Glass bottle
£36.88
5g
Please to purchase this item. Need a web account? Register with us today!
10483775
View Documents
Thermo Scientific Acros
192430250
25 g Glass bottle
£115.58
25g
Please to purchase this item. Need a web account? Register with us today!
10741535
View Documents
Thermo Scientific Acros
192431000
100 g Glass bottle
£337.68
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

10043-01-3
342.13
DIZPMCHEQGEION-UHFFFAOYSA-H
24850
dialuminum;trisulfate
Al2O12S3
MFCD00003423
aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
CHEBI:74768
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

770°C
White
15% min. (Al) Typical (Complexometry)
Al2(SO4)3
15, 360
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
342.13
CHEBI:74768
100.00%
Aluminum sulfate, Extra Pure
10043-01-3
Chunks and/or Powder
Al2O12S3
MFCD00003423
aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
DIZPMCHEQGEION-UHFFFAOYSA-H
dialuminum;trisulfate
24850
342.13
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-135-

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only