missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Sulfate Hexadecahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Specifications
Melting Point | 86°C |
Color | White |
pH | 2.5 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 342.1358g/mol |
Packaging | Plastic powder jar |
Safety and Handling

Product Identifier
- Aluminium sulfate hexadecahydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- H318-Causes serious eye damage.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Aluminum Sulfate Hexadecahydrate, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction