missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate octadecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

£40.50 - £116.58

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2O12S3·18H2O
Molecular Weight (g/mol) 666.4
MDL Number MFCD00149136
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10259500
View Documents
Thermo Scientific Acros
400595000
500 g Glass Bottle
£40.50
500g
Please to purchase this item. Need a web account? Register with us today!
10744124
View Documents
Thermo Scientific Acros
400590025
2.5 kg Plastic bottle
£116.58
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7784-31-8
666.4
AMVQGJHFDJVOOB-UHFFFAOYSA-H
22377415
dialuminum;trisulfate;octadecahydrate
Al2O12S3·18H2O
MFCD00149136
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
CHEBI:74779
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

86.0°C
White
98.0 to 102% (Complexometry)
Al2(SO4)3·18H2O
15, 360
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
666.4
CHEBI:74779
≥98%
Aluminum sulfate octadecahydrate
7784-31-8
Crystalline Powder
Al2O12S3·18H2O
MFCD00149136
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
AMVQGJHFDJVOOB-UHFFFAOYSA-H
dialuminum;trisulfate;octadecahydrate
22377415
666.4
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : WS5697000

Product Identifier
  • Aluminium sulfate octadecahydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only