Learn More
Fisherbrand™ Plane Swivel Scaffolding Connectors
Product Code. 11568112 Shop All Fisherbrand Products
Click to view available options
For Use With (Application):
Pivotal Plan 2-Way
Unit Size:
Each
Product Code. 11568112
Only null left
Add to basket
Add to basket
Suitable for making triangular or square frames for supporting large flanged items such as heating mantles and funnels. Fisherbrand™ Plane Swivel Scaffolding Connectors are provided with a steel strap.
  • With thumbscrews for hand tightening
  • For connecting two rods or tubes which converge at any angle
Specifications
Material Aluminum
Product Type Tripod Connection
For Use With (Application) Pivotal Plan 2-Way