missing translation for 'onlineSavingsMsg'
Learn More

Aluminon, ACS reagent

Product Code. 11945131 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11945131

Brand: Thermo Scientific Acros 401501000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 569-58-4 | C22H23N3O9 | 473.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-58-4
Molecular Formula C22H23N3O9
Molecular Weight (g/mol) 473.44
InChI Key AIPNSHNRCQOTRI-UHFFFAOYSA-N
Synonym aluminon, ammonium aurintricarboxylate, aurintricarboxylic acid ammonium salt, aurintricarboxylic acid, acs
PubChem CID 54729869
ChEBI CHEBI:87398
IUPAC Name triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]

Specifications

Melting Point 220.0°C to 225.0°C
Ignition Residue 0.2% max
Packaging Glass bottle
Quantity 100 g
Beilstein 10,II,775
Merck Index 15,318
Solubility Information Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol,insoluble in petroleum ether
Formula Weight 473.44
Grade ACS Reagent
Insoluble Matter 0.1% max
Chemical Name or Material Aluminon
Show More Show Less
What is the solubility of Aluminon?

Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.

compliance-icons
Product Identifier
  • Aluminon
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Aluminon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.