Learn More
Fisherbrand™ Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Bossheads come with offset jaws for greater strength.
Brand: Fisherbrand™
Code : 41
Additional Details : Weight : 0.29520kg
Beschreibung
- With offset knobs for greater strength
- Aluminum alloy with clamping screws
- For use with rods up to 16mm in diameter
- Note: these screws have a metal head
Spezifikation
5/Pk. | |
Aluminum Alloy | |
For use with rod up to 16mm diameter |
Bosshead Clamp | |
Metal headed clamping screw |