missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Bossheads come with offset jaws for greater strength.
£6.74 - £106.00
Specifications
Quantity | 5/Pk. |
---|---|
Type | Bosshead Clamp |
Material | Aluminum Alloy |
For Use With (Equipment) | For use with rod up to 16mm diameter |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11537722
|
Fisherbrand™
|
Metal headed clamping screw |
£106.00
Pack of 5 |
In Stock
Log in to see stock. |
|||||
11547722
|
Fisherbrand™
|
Plastic headed clamping screws |
£105.26
Pack of 5 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
10758931
|
Fisherbrand™
|
Metal headed clamping screw |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Aluminum alloy with clamping screws
- For use with rods up to 16mm in diameter
Specifications
5/Pk. | |
Bosshead Clamp | |
Aluminum Alloy | |
For use with rod up to 16mm diameter |