missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ammonium sulfate dodecahydrate, Reagent Grade, Thermo Scientific Chemicals

£24.22 - £150.75

Chemical Identifiers

CAS 7784-26-1
Molecular Formula AlH28NO20S2
Molecular Weight (g/mol) 453.313
MDL Number MFCD00051042
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
PubChem CID 62668
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361349
View Documents
Thermo Scientific Alfa Aesar
013802.22
100 g
£24.22
100g
Please to purchase this item. Need a web account? Register with us today!
11351349
View Documents
Thermo Scientific Alfa Aesar
013802.36
500 g
£54.67
500g
Please to purchase this item. Need a web account? Register with us today!
11341349
View Documents
Thermo Scientific Alfa Aesar
013802.A3
2 kg
£150.75
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Water and sewage purification, dyeing and printing fabrics, manufacturing of pigments, paper, artificial gems, cements, in electrolytic copper platingAluminum ammonium sulfate dodecahydrate acts as a crystalline double sulfate. It is used in water purification, in vegetable glues, in porcelain cements, in deodorant, in tanning and in fireproofing textiles. It serves as an ingredient in animal repellant sprays. Further, it is used in the preparation of Alum (Al(OH)3) gel. In addition, it is used in cosmetic formulation, fertilizers, food preservation and in the heating system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7784-26-1
453.313
WZUKKIPWIPZMAS-UHFFFAOYSA-K
62668
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
AlH28NO20S2
MFCD00051042
ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
aluminum;azanium;disulfate;dodecahydrate

Specifications

93°C to 94°C
White
AlH28NO20S2
ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
WZUKKIPWIPZMAS-UHFFFAOYSA-K
aluminum;azanium;disulfate;dodecahydrate
62668
Reagent
Aluminum ammonium sulfate dodecahydrate
7784-26-1
Crystals
MFCD00051042
Soluble in water and glycerol. Insoluble in alcohol.
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
453.313
453.33 (237.14 Anhydrous)
1.65
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 232-055-3

missing translation for 'rtecsNumber' : WS5640010

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Aluminum ammonium sulfate dodecahydrate

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only