missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wesbart™ Aluminum Anodized Test Tube Racks
Wesbart aluminum anodized racks to store test tubes
Brand: Wesbart™ SP2050
This item is not returnable.
View return policy
Description
- Single, double, or triple platform
- aluminum test tube racks to store tubes.
Specifications
Test Tube Rack | |
Autoclavable | |
50 | |
Yes | |
140 x 283 x 35 mm | |
20 mm |
Aluminum Test Tube Rack | |
Tubes (20 mm) | |
Rectangular | |
Non-sterile | |
35 mm |