missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Aluminum Basin with Lid

Made of durable aluminum, these Nickel Electro™ Aluminum Basins with Lid  are ideal in laboratory environments.

Brand:  Nickel Electro™ 650

Code : 03

Additional Details : Weight : 0.02340kg

 View more versions of this product

Product Code. 12747356

  • £36.70 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • For coal moisture determination
  • Flat, bottom aluminum basin with lid 
Specifications

Specifications

Flat
Aluminum
For Coal Moisture Determination
65 mm
45 mL
Evaporating Basin
14 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers