Learn More
Aluminum bromide, 98%, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 011113.59
Code : NEW
Additional Details : CAS Number : 7727-15-3 Weight : 0.50000kg Transport : UN number : 1725 Chem class : 8 Pack group : II
Description
Alkylation, bromination and isomerization catalyst in organic synthesis.Aluminum bromide acts as a strong Lewis acid and is used in Lewis acid-promoted reactions such as epoxide ring openings. It is used as a catalyst for Friedel-Crafts alkylation reaction. It also finds application in bromination and isomerization reactions in organic synthesis. It is also used in polymerization reaction to prepare poly(o-xylene)from benzocyclobutene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation, bromination and isomerization catalyst in organic synthesisSpecifications
265°C | |
7727-15-3 | |
5 x 100 g | |
Aluminum tribromide; |
97.5°C | |
Solid | |
AlBr{3} | |
98% |