missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Aluminum Ceramic Alsint Flat Bottom Crucible

Alsint 99.7 crucibles are an excellent choice for use under high-wear conditions, in chemical technical and electrical engineering applications and in high-temperature technology.

Brand:  Haldenwanger™ L ALSINTTIEGEL*A/7

Product Code. 10292161

  • £545.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High refractoriness (melting point above 2000°C)
  • Chemical resistance to a wide range of substances
  • Applications include crystal growing, fusion processes, annealing
  • Excellent for use with ball mill pots, milling balls, mouthpieces, shaft-protection sleeves
  • Cylindrical shape, flat base
Specifications

Specifications

White
Aluminum Ceramic
85 mm
Alsint Crucible
75 mm
700 mL
150 mm
Cylinder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers