Learn More
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible
Use these crucibles for general laboratory applications. Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucibles are resistant to a variety of acids and alkalis.
Brand: Haldenwanger™ L ALSINTTIEGEL*B/3
This item is not returnable.
View return policy
Description
- Conical squat form crucibles
- Aluminum ceramic composition
- Available in a variety of sizes
- Aluminum ceramic composition
Specifications
Aluminum Ceramic | |
80 mL | |
Crucible | |
Conical |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible > Capacity: 80mL
Spot an opportunity for improvement?Share a Content Correction