Learn More
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible
Product Code. 10493711
Click to view available options
Capacity (Metric):
10 mL
25 mL
40 mL
60 mL
80 mL
Unit Size:
Each
Product Code. 10493711
Brand: Haldenwanger™ L ALSINTTIEGEL*B/1
Only null left
Add to basket
Add to basket
Use these crucibles for general laboratory applications. Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucibles are resistant to a variety of acids and alkalis.
  • Conical squat form crucibles
  • Aluminum ceramic composition
  • Available in a variety of sizes
  • Aluminum ceramic composition
Specifications
Material Aluminum Ceramic
Capacity (Metric) 40 mL
Product Type Crucible
Shape Conical