Learn More
Bochem™ Aluminum Chemical Scoop
Manufactured from aluminum, the Bochem™ Aluminum Chemical Scoop – is a perfect addition to your laboratory tools.
£2.15 - £48.95
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10655991
|
Bochem™
8821 |
400 mL | 290 mm |
£11.85
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
10360341
|
Bochem™
8815 |
10 mL | 52 mm |
£2.15
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
||||
10796971
|
Bochem™
8817 |
65 mL | 155 mm |
£4.90
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
10122221
|
Bochem™
8824 |
1.6 L | 270 mm |
£48.95
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
10554841
|
Bochem™
8827 |
90 mL | 250 mm |
£12.30
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
||||
10652882
|
Bochem™
8822 |
480 mL | 310 mm |
£13.55
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
10662882
|
Bochem™
8823 |
900 mL | 350 mm |
£28.25
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
10603272
|
Bochem™
8820 |
240 mL | 145 mm |
£7.75
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
10386001
|
Bochem™
8828 |
90 mL | 285 mm |
£10.15
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
Description
- Lightweight
- Aluminum
- Length is total for combined scoop and handle
Specifications
Chemical Scoop | |
Aluminum |