missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Aluminum Chemical Scoops
Manufactured from aluminum. Bochem™ Aluminum Chemical Scoops comes in many sizes.
Brand: Bochem™ 8819
Code : A0
Additional Details : Weight : 0.50000kg
Description
- Chemical scoop
- Manufactured from aluminum
- Comes in many sizes
Specifications
1 Ea. | |
Chemical Scoop | |
Aluminum |
160 mL | |
37mm | |
210 mm |