missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, 0.5M aq. soln., Thermo Scientific™

£23.81 - £49.45

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15474159
SDS View Documents
Thermo Scientific Alfa Aesar
J61582.AD
50 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15484159
SDS View Documents
Thermo Scientific Alfa Aesar
J61582.AE
100 mL
£49.45
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7784-13-6
241.42
JGDITNMASUZKPW-UHFFFAOYSA-K
AlCl3H12O6
MFCD00149134
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

7784-13-6
Liquid
MFCD00149134
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminum(3+) hexahydrate trichloride
133.34
Aluminum chloride
Colorless
AlCl3H12O6
UN2581
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
241.42
0.5M aq. soln.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALUMINUM CHLORIDE, SOLUTION

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum chloride, 0.5M aq. soln., Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.