missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, anhydrous, 99.985% (metals basis), Thermo Scientific Chemicals

Catalog Number 11359279
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy
7446-70-0
AlCl3
133.33
MFCD00003422
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
aluminum(3+) trichloride
[Al+3].[Cl-].[Cl-].[Cl-]
This item is not returnable. View return policy
7446-70-0
AlCl3
133.33
MFCD00003422
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
aluminum(3+) trichloride
[Al+3].[Cl-].[Cl-].[Cl-]

Lithium sulfate, anhydrous is used to treat bipolar disorder, solar panels and the potential for a new class of battery, and used as a catalyst for the elimination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY
Melting Point 190° (sublimation)
Color Gray to Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.985% (Metal basis)
Solubility Information Soluble in water,hydrochloric acid and ethanol. Slightly soluble in benzene.
Formula Weight 133.34
Odor Pungent
Sensitivity Moisture sensitive
Appearance Yellow to gray
Density 2.44 g/cm3
Chemical Name or Material Aluminum chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'