missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride hexahydrate, 99%

Código de producto. 11497077
Click to view available options
Quantity:
250 g
1000 g
5000 g
Tamaño de la unidad:
1000g
250g
5000g
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11497077

Marca: Thermo Scientific Alfa Aesar A14437.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 7784-13-6 | AlCl3H12O6 | 241.42 g/mol

Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

Solubility
Highly soluble in water. Soluble in alcohol, ether, glycerol, propylene glycol and acetonitrile.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Especificaciones

Melting Point 100°C (decomposition)
pH 2.5
Quantity 1000 g
UN Number UN3260
Merck Index 14,337
Solubility Information Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile.
IUPAC Name aluminum(3+) hexahydrate trichloride
Formula Weight 241.43 (133.34 Anhydrous)
Percent Purity 99%
Odor Odorless
Density 2.398
Chemical Name or Material Aluminum chloride hexahydrate
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Aluminum chloride hexahydrate
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Aluminium chloride,hexahydrate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado