missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum chloride, Anhydrous, sublimed, ≥98%, Honeywell Fluka™
Anhydrous, sublimed,≥98%
£29.50 - £161.40
Chemical Identifiers
CAS | 7446-70-0 |
---|---|
Molecular Formula | AlCl3 |
Molecular Weight (g/mol) | 133.33 |
MDL Number | MFCD00003422 |
InChI Key | VSCWAEJMTAWNJL-UHFFFAOYSA-K |
Synonym | aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum |
IUPAC Name | aluminum(3+) trichloride |
SMILES | [Al+3].[Cl-].[Cl-].[Cl-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7446-70-0 | |
133.33 | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride |
AlCl3 | |
MFCD00003422 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
[Al+3].[Cl-].[Cl-].[Cl-] |
Specifications
7446-70-0 | |
AlCl3 | |
MFCD00003422 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
[Al+3].[Cl-].[Cl-].[Cl-] | |
133.33 | |
≥98% | |
Aluminum Chloride |
Anhydrous, Sublimed | |
AlCl3 | |
1726 | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride | |
133.34g/mol | |
1mmHg (100°C) |
Safety and Handling
P280-P305 + P351 + P338-P310
H314-H372-H373
missing translation for 'einecsNumber' : 231-208-1
missing translation for 'rtecsNumber' : BD0525000