missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 12 x 32mm Vials

You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.

Brand:  Thermo Scientific™ C4011-5A

 View more versions of this product

Product Code. 12413778

This item is not returnable. View return policy

Description

Description

You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.

  • Anodized aluminum seals fit all 11mm crimp-top vials
  • Seal 2000™ material is specially formulated for improved background performance over PTFE/red rubber
  • Viton™ septa perform well with ECD detectors and are resistant to many aggressive solvents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black Viton Elastomer Septa; Silver Aluminum Seal
Viton Elastomer, Aluminum
National
11 mm
Silver, Black
Black Viton Elastomer
Crimp Cap
12x32 Vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers