missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 12 x 32mm Vials

You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.

Brand:  Thermo Scientific™ C4011-4Y

 View more versions of this product

Product Code. 12443738

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.

  • Anodized aluminum seals fit all 11mm crimp-top vials
  • Seal 2000™ material is specially formulated for improved background performance over PTFE/red rubber
  • Viton™ septa perform well with ECD detectors and are resistant to many aggressive solvents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prefitted Red PTFE/White Silicone Septa; Yellow Aluminum Seal; Autosampler
PTFE, Silicone, Aluminum
Crimp Cap
12x32 Vials
Yellow, Red, White
Prefitted Red PTFE/White Silicone Septa
11 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers