missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Aluminum di(isopropoxide)acetoacetic ester chelate is used as a gel ink. It finds application in paint to give better drying, luster and hardness, and improve the performance as surface wrinkling. It is also used as a crosslinking agent in epoxy, alkyd and polyester resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.05 g/mL |
Flash Point | 50°C (122°F) |
Odor | Alcohol-like |
Quantity | 100 g |
UN Number | UN1993 |
Sensitivity | Moisture sensitive |
Formula Weight | 274.3 |
Percent Purity | ≥9.6% |
Physical Form | Liquid |
Chemical Name or Material | Aluminum di (isopropoxide)acetoacetic ester chelate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'