Learn More
Aluminum dihydrogen phosphate, 50% w/w aqueous solution, Thermo Scientific Chemicals
£88.24 - £298.49
Chemical Identifiers
CAS | 13530-50-2 |
---|---|
Molecular Formula | AlH6O12P3 |
Molecular Weight (g/mol) | 317.94 |
MDL Number | MFCD00014152 |
InChI Key | RGPUVZXXZFNFBF-UHFFFAOYSA-K |
Synonym | aluminum dihydrogen phosphate, aluminum phosphate monobasic, prim-aluminum phosphate, aluminum tris dihydrogen phosphate, phosphoric acid, aluminum salt 3:1, aluminum hydrogen phosphate, aluminumdihydrogen phosphate, aluminum phosphate,monobasic, aluminum trisdihydrogenphosphate, aluminum dihydrogen orthophosphate |
PubChem CID | 9818426 |
IUPAC Name | aluminum;dihydrogen phosphate |
SMILES | [Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11310098
|
Thermo Scientific Alfa Aesar
042858.A1 |
1 kg |
£88.24
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11300098
|
Thermo Scientific Alfa Aesar
042858.A7 |
5 kg |
£298.49
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum dihydrogen phosphate is used as intermediates and solid separation agents. It is also used in petroleum, chemical industry, shipbuilding, space technology, electric industry, high-temperature kilns, heat treatment resistance furnaces and electric insulation. Further, it is used as inorganic coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13530-50-2 | |
317.94 | |
RGPUVZXXZFNFBF-UHFFFAOYSA-K | |
9818426 | |
[Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O |
AlH6O12P3 | |
MFCD00014152 | |
aluminum dihydrogen phosphate, aluminum phosphate monobasic, prim-aluminum phosphate, aluminum tris dihydrogen phosphate, phosphoric acid, aluminum salt 3:1, aluminum hydrogen phosphate, aluminumdihydrogen phosphate, aluminum phosphate,monobasic, aluminum trisdihydrogenphosphate, aluminum dihydrogen orthophosphate | |
aluminum;dihydrogen phosphate |
Specifications
13530-50-2 | |
Liquid | |
AlH6O12P3 | |
aluminum dihydrogen phosphate, aluminum phosphate monobasic, prim-aluminum phosphate, aluminum tris dihydrogen phosphate, phosphoric acid, aluminum salt 3:1, aluminum hydrogen phosphate, aluminumdihydrogen phosphate, aluminum phosphate,monobasic, aluminum trisdihydrogenphosphate, aluminum dihydrogen orthophosphate | |
RGPUVZXXZFNFBF-UHFFFAOYSA-K | |
aluminum;dihydrogen phosphate | |
9818426 | |
Colorless liquid | |
Aluminum dihydrogen phosphate |
Colorless | |
50% w/w aqueous solution | |
MFCD00014152 | |
Soluble in water and ethanol. Insoluble in acetone. | |
[Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O | |
317.94 | |
317.94 | |
1.5 |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum dihydrogen phosphate, 50% w/w aq. Soln
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Aluminum dihydrogenphosphate
RUO – Research Use Only