missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Aluminum Dish and Cover, Flat Bottom and Straight Sides
£15.97 - £115.00
Product Code | Brand | Diameter (Metric) | Height (Metric) | No. per Pack | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | No. per Pack | Price | Quantity & Availability | |||
11470201
|
Thermo Scientific™
039077.KH |
51 mm | 12.5 mm | 2/Pk. | |||||
11303468
|
Thermo Scientific™
039077.UD |
51 mm | 12.5 mm | 6/Pk. | |||||
11490201
|
Thermo Scientific™
039079.KH |
63 mm | 44 mm | 2/Pk. | |||||
11323468
|
Thermo Scientific™
039079.UD |
63 mm | 44 mm | 6/Pk. | |||||
11400211
|
Thermo Scientific™
039080.KH |
76 mm | 25 mm | 2/Pk. | |||||
11333468
|
Thermo Scientific™
039080.UD |
76 mm | 25 mm | 6/Pk. | |||||
11430211
|
Thermo Scientific™
039083.KH |
204 mm | 25 mm | 2/Pk. | |||||
11440211
|
Thermo Scientific™
039084.KH |
204 mm | 51 mm | 2/Pk. | |||||
11363468
|
Thermo Scientific™
039083.UD |
204 mm | 25 mm | 6/Pk. | |||||
11373468
|
Thermo Scientific™
039084.UD |
204 mm | 51 mm | 6/Pk. | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Aluminum |