missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Aluminum Fixed Angle Rotors

Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.

Brand:  Eppendorf™ 5424702007

630.00 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10555563

  • £677.70 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Choice of four different rotors
  • Standard rotor can accommodate 1.5mL/2.0mL tubes
  • PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aluminum
24 x 1.5/2 mL
Aerosol rotor lid
Fixed Angle Rotor
14,680 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.