Learn More
Aluminum flake, APS 11 micron, 99.7% (metals basis), Thermo Scientific Chemicals
£37.00 - £375.87
Chemical Identifiers
CAS | 7429-90-5 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
SMILES | [Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446685
|
Thermo Scientific Alfa Aesar
045031.30 |
250 g |
£37.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456685
|
Thermo Scientific Alfa Aesar
045031.A1 |
1 kg |
£90.35
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15466685
|
Thermo Scientific Alfa Aesar
045031.A7 |
5 kg |
£375.87
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
[Al] |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminumShow More | |
CHEBI:33629 |
Specifications
Silver to Gray | |
Silver Gray flakes | |
99.7% (Metals basis) | |
MFCD00134029 | |
400°C (752°F) | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Water reactive | |
Aluminum |
7429-90-5 | |
2.7g/cm3 | |
Al | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminum | |
5359268 | |
Odorless | |
Flake |
Safety and Handling
Keep container tightly closed and dry.
Contact with water liberates highly flammable gases. Spontaneously flammable in air.
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II
missing translation for 'einecsNumber' : 231-072-3
missing translation for 'rtecsNumber' : BD0330000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Aluminum flake
Signal Word
- Danger
Hazard Category
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P370+P378-In case of fire: Use to extinguish.
- P422-Store contents under .
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.