missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Aluminum Block, Flat Bottom, for 96/384 Well Assay Plates
Click to view available options
Length (Metric):
39.2 mm
Description
- 1 year warranty
Specifications
Specifications
Holds | 384-well Assay Plates |
Material | Aluminum |
No. of Places | 96 |
Type | Block |
Length (English) | 1.5 in. |
Length (Metric) | 39.2 mm |
Width (English) | 1.4 in. |
Width (Metric) | 35.6 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'