missing translation for 'onlineSavingsMsg'
Learn More
Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11300809

Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11300809
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11300809

Brand: Thermo Scientific Alfa Aesar 010623.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32287-65-3
Molecular Formula AlF3H2O
Molecular Weight (g/mol) 101.99
MDL Number MFCD00149142
InChI Key XTXFDUABTPNTFB-UHFFFAOYSA-K
Synonym aluminum fluoride hydrate, alf3.h2o, trifluoroalumane-water 1/1, aluminum fluoride hydrate, puratronic
PubChem CID 16211462
IUPAC Name trifluoroalumane hydrate
SMILES O.F[Al](F)F

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN3260
Formula Weight 83.98 (Anhydrous)
Percent Purity 99.99%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Aluminum fluoride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.