missing translation for 'onlineSavingsMsg'
Learn More

Aluminum fluoride trihydrate, 97%, Thermo Scientific Chemicals

£29.15 - £245.22

Chemical Identifiers

CAS 15098-87-0
Molecular Formula AlF3H6O3
Molecular Weight (g/mol) 138.022
MDL Number MFCD00003426
InChI Key BKLZIAYVINRQEJ-UHFFFAOYSA-K
Synonym aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
PubChem CID 22002992
IUPAC Name aluminum;trifluoride;trihydrate
SMILES O.O.O.[F-].[F-].[F-].[Al+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347167
View Documents
Thermo Scientific Alfa Aesar
011496.22
100 g
£29.15
100g
Please to purchase this item. Need a web account? Register with us today!
11357167
View Documents
Thermo Scientific Alfa Aesar
011496.36
500 g
£84.12
500g
Please to purchase this item. Need a web account? Register with us today!
11367167
View Documents
Thermo Scientific Alfa Aesar
011496.A3
2 kg
£245.22
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15098-87-0
138.022
BKLZIAYVINRQEJ-UHFFFAOYSA-K
22002992
O.O.O.[F-].[F-].[F-].[Al+3]
AlF3H6O3
MFCD00003426
aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
aluminum;trifluoride;trihydrate

Specifications

250°C (in -3H2O)
White
AlF3H6O3
14,339
Slightly soluble in water,acids and alkalis. Insoluble in alcohol and acetone.
O.O.O.[F-].[F-].[F-].[Al+3]
138.022
138.02 (83.98 Anhydrous)
Odorless
Aluminum fluoride trihydrate
15098-87-0
Powder
MFCD00003426
aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
BKLZIAYVINRQEJ-UHFFFAOYSA-K
aluminum;trifluoride;trihydrate
22002992
97%
2.17 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H320-H335

missing translation for 'einecsNumber' : 232-051-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Aluminum fluoride trihydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only