missing translation for 'onlineSavingsMsg'
Learn More
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11470700

Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11470700
50 x 50 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11470700

Brand: Thermo Scientific Alfa Aesar 044331.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Form: Approx. 1.7g/50 x 50mm; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored foil
Assay Percent Range 99.9999% (Metals basis)
Quantity 50 x 50 mm
IUPAC Name aluminum
Odor Odorless
Form Foil
Chemical Name or Material Aluminum, Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR10I013 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog Number044331.FI,044331.HP,044331.GH
Lot NumberQ27I055 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog Number044331.FI,044331.HP,044331.GH
Lot NumberJ25W040 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044331.FI,044331.HP,044331.GH
Lot NumberL09Q015 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044331.FI,044331.HP,044331.GH
Lot NumberC25X034 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044331.FI,044331.HP,044331.GH

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.