Aluminum foil, 0.5mm (0.02in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals
Product Code.: 11470281
Product Code. 11470281
50 x 50 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code.
11470281
Brand: Thermo Scientific Alfa Aesar
040531.FI
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
7429-90-5
Molecular Formula
Al
Molecular Weight (g/mol)
26.98
MDL Number
MFCD00134029
InChI Key
XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID
5359268
ChEBI
CHEBI:33629
IUPAC Name
aluminum
SMILES
[Al]
Specifications
Quantity
50 x 50 mm
Percent Purity
99.99%
Odor
Odorless
Weight
≈3.4g/50x50mm
Form
Foil
Assay
(metals basis)
Chemical Name or Material
Aluminum foil, 0.5mm (0.02 in.) thick, Annealed, Temper: soft
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number
Certificate Type
Certificate of Analysis
Certificate of Compliance
Certificate of Sterility/Irradiation
Certificate of Validation
Certificate of Guarantee
Certificate of Animal Origin
Certificate of Quality
Certificate of Processing
Declaration of Conformity
Search
5 results found
Lot Number
Certificate Type
Date
Catalog Number
Lot Number Y02H003
Certificate Type Certificate of Analysis
Date 12/04/2022
Catalog Number 040531.FI,040531.GH,040531.HP
Lot Number E23X045
Certificate Type Certificate of Analysis
Date 07/12/2021
Catalog Number 040531.FI,040531.GH,040531.HP
Lot Number F25M14
Certificate Type Certificate of Analysis
Date 07/12/2021
Catalog Number 040531.FI,040531.GH,040531.HP
Lot Number F09U030
Certificate Type Certificate of Analysis
Date 07/12/2021
Catalog Number 040531.FI,040531.GH,040531.HP
Lot Number T15H033
Certificate Type Certificate of Analysis
Date 07/12/2021
Catalog Number 040531.FI,040531.GH,040531.HP
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Aluminum foil, 0.5mm (0.02in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.