missing translation for 'onlineSavingsMsg'
Learn More
Aluminum foil, 1.0mm (0.04in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11355067

Aluminum foil, 1.0mm (0.04in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11355067
100 x 100 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
This item is not returnable. View return policy

Product Code. 11355067

Brand: Thermo Scientific Alfa Aesar 040762.GH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 100 x 100 mm
Percent Purity 99.99%
Odor Odorless
Weight ≈6.75g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Aluminum foil, 1.0mm (0.04 in.) thick, Annealed, Temper: soft

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW27D005 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog Number040762.HP,040762.FI,040762.GH
Lot NumberN10I062 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog Number040762.HP,040762.FI,040762.GH
Lot NumberW19A030 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040762.HP,040762.FI,040762.GH
Lot NumberF08W011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040762.HP,040762.FI,040762.GH
Lot NumberD13X016 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040762.HP,040762.FI,040762.GH

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum foil, 1.0mm (0.04in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.