missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum foil, 1.5mm (0.06 in.) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%, Thermo Scientific™
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Quantity | 50 x 50 mm |
Percent Purity | 99% |
Concentration or Composition (by Analyte or Components) | Si and Fe (combined) typically 1% |
Odor | Odorless |
Weight | ≈10.125g/50x50mm |
Form | Foil |
Assay | (metals basis) |
Chemical Name or Material | Aluminum foil, 1.5mm (0.06 in.) thick, Annealed, Temper: soft |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'