Learn More
Bryson Packaging™ Aluminum Foil Rolls
Ideal for use in autoclaves. Bryson Packaging™ Aluminum Foil Rolls are suitable for many laboratory applications including sealing bottles, culture tubes, stopping leaks, reducing evaporation or heat losses, covering corks and making weighing boats or dishes.
Brand:  Bryson Packaging™ AKL-300-030M
 View more versions of this product
Product Code. 12369548
  • £157.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Available in a variety of widths
  • Can be cut to any length, cutter provided with box
  • Sealing, weighing and transferring samples
Specifications
Specifications
Ideal for use in autoclaves; 'Laboratory applications -including sealing bottles, culture tubes, stopping leaks, reducing evaporation or heat losses, covering corks and making weighing boats or dishes
450mm
12 μm
Foil
Aluminum
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bryson Packaging™ Aluminum Foil Rolls > Dimensions (L x W): 75m x 450mm