Learn More
Wesbart™ Aluminum Horizontal Freezer Rack
Wesbart™ Aluminum Horizontal Freezer Racks are front loading with trays for quick access to cryoboxes and other stored items.
£86.10 - £245.00
Product Code | Brand | Holds | Positions | Dimensions (L x W x H) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Positions | Dimensions (L x W x H) | Price | Quantity & Availability | |||
15726418
|
Wesbart™
AH179/3UT |
Boxes (12/Rack) | 3 Trays/Rack | 560 x 140 x 420 mm |
£123.00
Each |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
15951231
|
Wesbart™
AH506/2AT |
Boxes (4/Rack) | 2 Trays/Rack | 560 x 140 x 235 mm |
£95.65
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
15905590
|
Wesbart™
BH115/5NMT |
Boxes (25/Rack) | 5 Trays/Rack | 685 x 140 x 287 mm |
£209.50
Each |
In Stock
Log in to see stock. |
|||
16224941
|
Wesbart™
BH115/2NMT |
Boxes (10/Rack) | 2 Trays/Rack | 685 x 140 x 287 mm |
£106.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15768666
|
Wesbart™
AH179/7UT |
Boxes (28/Rack) | 7 Trays/Rack | 560 x 140 x 415 mm |
£245.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
15710727
|
Wesbart™
AH179/7U |
Boxes (28/Rack) | 7 Trays/Rack | 560 x 140 x 420 mm |
£134.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
15884732
|
Wesbart™
HC2706 |
Loose Items or Cryoboxes | 1 Tray/Rack | 560 x 140 x 290 mm |
£111.38
Each |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16265970
|
Wesbart™
BH195/2NMT |
Loose Items or Cryoboxes | 2 Trays/Rack | 675 x 155 x 290 mm |
£134.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||
15897642
|
Wesbart™
BH174/2NMT |
Loose Items or Cryoboxes | 2 Trays/Rack | 688 x 140 x 300 mm |
£109.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||
15893975
|
Wesbart™
HCS46/4 |
Loose Items or Cryoboxes | 4 Trays/Rack (Not Included) | 560 x 145 x 235 mm |
£97.80
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||
15988496
|
Wesbart™
AH900/4 |
Loose Items or Cryoboxes | 4 Trays/Rack | 640 x 165 x 240 mm |
£86.10
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
Description
- Aluminum front access storage rack for tray units (ordered separately, Cat. No. 17311849) for upright freezers
- For standard cryobox or other freezer storage
- Only one tray has to be removed from the rack at a time for sample access, minimizing temperature impact on remaining items
- Range of sizes and capacities available